Vogel-Johnson Agar (Dehydrated)
Thermo Scientific™

Vogel-Johnson Agar (Dehydrated)

Isolate and presumptively identify Staphylococcus aureus from clinical specimens and food with this selective medium.

Have Questions?
Catalog NumberQuantity
CM0641B500 g
Catalog number CM0641B
Price (INR)
-
Quantity:
500 g

Isolate and presumptively identify Staphylococcus aureus from clinical specimens and food with Thermo Scientific™ Oxoid™ Vogel-Johnson Agar (Dehydrated). The medium is based on the formula corresponding to the United States Pharmacopoeia1. Staphylococcus aureus reduces tellurite to metallic tellurium in the medium and appear as black colonies.

Staphylococcus aureus is a Gram-positive, round-shaped bacteria and can cause skin and soft tissue infections such as abscesses (boils), furuncles, and cellulitis. Although most staph infections are not serious, S. aureus can cause serious infections such as bloodstream infections, pneumonia, or bone and joint infections.

Vogel-Johnson Agar allows the early detection and of Staphylococcus aureus from heavily contaminated foods and clinical specimens by selecting and identifying coagulase-positive and monitol-fermenting strains.

  • Selective: tellurite, lithium chloride and the high glycine concentration inhibits most of the contaminating organisms.
  • Easy-to-read: Staphylococcus aureus appear as black colonies due to their ability to reduce tellurite to metallic tellurium.
  • Conforms to the United States Pharmacopoeia1.
  • Allows the early detection and of Staphylococcus aureus from heavily contaminated foods and clinical specimens by selecting and identifying coagulase-positive and monitol-fermenting strains.

Vogel and Johnson2 modified the Tellurite Glycine Agar formula of Zebovitz et al.3 by doubling the mannitol concentration to 1% (w/v) and adding Phenol Red as a pH indicator. The enhanced fermentation reaction which occurs as a result of the increase in mannitol content is clearly indicated by the development of yellow zones surrounding the colonies.

Prolonged incubation may result in the growth of black coagulase-negative colonies and if these organisms also ferment mannitol they may be falsely identified from their appearance as Staphylococcus aureus. In these circumstances, further tests of identity should be carried out before concluding that the organism is Staphylococcus aureus.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For Laboratory Use Only

General References:

  1. United States Pharmacopoeia XXI (1985) Microbial. Limit Tests. Rockville. Md.
  2. Vogel R. A. and Johnson M. J. (1961) Pub. Hlth Lab. 18. 131.
  3. Zeboritz E., Evans J. B. and Niven C. F. (1955) J. Bact. 70. 687.
Specifications
Certifications/ComplianceIndustrial Reference: FDA.
DescriptionVogel-Johnson Agar
Quantity500 g
YieldFor 8.2L medium
FormPowder
Product TypeAgar
Unit SizeEach