Haemophilus Test Medium Base (Dehydrated)
Thermo Scientific™
Haemophilus Test Medium Base (Dehydrated)
Carry out susceptibility testing of Haemophilus influenzae with highly reproducible results with this medium.
Have Questions?
Catalog NumberQuantity
CM0898B500 g
Catalog number CM0898B
Price (INR)
-
Quantity:
500 g
Carry out susceptibility testing of Haemophilus influenzae with Thermo Scientific™ Oxoid™ Haemophilus Test Medium Base (Dehydrated). The medium is based on the formulation developed by Jorgensen et al1 which is now recommended by United States CLSI (M100Ed 29). The results achieved using Haemophilus Test Medium Base are highly reproducible3,4.
Haemophilus species are nutritionally fastidious in nature and require complex media for growth. Because of the antagonism between some essential nutrients and certain antimicrobial agents, the complex media formulations make routine susceptibility testing of Haemophilus species difficult. Regular Mueller Hinton Agar requires the addition of blood to supply essential nutrients making medium opaque, rendering it unsuitable for antimicrobial susceptibility testing.
Haemophilus Test Medium is based on the formulation developed by Jorgensen et al.2 which is now recommended by the United States CLSI. The transparency of the medium allows zones of inhibition to be read easily through the bottom of the Petri dish. Haemophilus Test Medium Base contains low levels of antimicrobial antagonists, which allows testing of trimethoprim/sulphamethoxazole to be carried out with greater confidence.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
General References:
  1. Jorgensen J.H., Redding J.S., Maher L.A. and Howell A.W. (1987) J. Clin. Micro. 25, 2105-2113.
  2. CLSI M100 ed29.
  3. Doern G.V., Jorgensen J.H., Thornsberry C. and Snapper H. (1990). Eur. J. Clin. Microbiol. Infect. Dis. 9, 329-336.
  4. Barry A.L., Jorgensen J.H. and Hardy D.J. (1991) J. Antimic. Chem. 27, 295-301.
Specifications
DescriptionHaemophilus Test Medium Base
Quantity500 g
YieldFor 11.6L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach