Brilliance™ Candida Agar Base (Dehydrated)
Thermo Scientific™
Brilliance™ Candida Agar Base (Dehydrated)
Rapidly isolate and identify clinically important Candida species with this medium.
Have Questions?
Catalog NumberQuantity
CM1002B500 g
Catalog number CM1002B
Price (INR)
-
Quantity:
500 g
Rapidly isolate and identify clinically important Candida species with Thermo Scientific™ Brilliance™ Candida Agar Base (Dehydrated). This chromogenic formulation produces four different diagnostic colors for the presumptive identification of clinically significant Candida species and provides faster, easier-to-read results when compared to traditional media.
Serious infections due to Candida species are becoming increasingly prevalent. This poses particular problems because of the increasing incidence of non-albicans spp. and the emergence of non-albicans isolates resistant to both amphotericin B and the newer azoles.1
Isolate, differentiate and identify Candida albicans and Candida tropicalis from other Candida spp. within 48 hours with this selective differential chromogenic medium.
  • Rapid: allows the identification of Candida albicans and Candida tropicalis from other species of Candida within 48 hours.
  • Easy-to-read:Candidaalbicans (green colonies) is easy to isolate from Candida tropicalis (dark blue colonies)
Brilliance Candida Agar incorporates two chromogens that indicate the presence of the target enzymes:
  1. X-NAG (5-bromo-4-chloro-3-indolyl N acetyl ß-D-glucosaminide) detects the activity of hexosaminidase.
  2. BCIP (5-bromo-6-chloro-3-indolyl phosphate p-toluidine salt) detects alkaline phosphatase activity
The typical enzyme patterns of Candida spp. as below:
Chromogen:
Enzyme:
X-NAG
Hexosaminidase
BCIP
Alkaline phosphatase
Typical colony appearance
C. tropicalis+ Dark blue
C. albicans
C. dubliniensis
+ Green ‡
C. krusei +Dry, irregular pink-brown
C. glabrata
C. kefyr
C. parapsilosis
C. lusitaniae
 VariableBeige/yellow/ brown †
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
1. Sheehan, D. J. et al. (1999) Current and Emerging Azole Antifungal Agents. Clinical Microbiology Reviews, 12(1): 40-79.
Specifications
DescriptionBrilliance Candida Agar Base
Quantity500 g
YieldFor 16.0L medium
FormPowder
Product TypeAgar
Unit SizeEach