Bax Listeria Medium
Thermo Scientific™
Bax Listeria Medium
Detect Listeria with the BAX® System using this broth.
Have Questions?
Catalog NumberQuantity
CM1067R2.5 kg
Catalog number CM1067R
Price (INR)
-
Quantity:
2.5 kg
Bax Listeria Medium is a single selective enrichment broth optimized to detect Listeria with the BAX® System
Specifications
FormatPowder
Quantity2.5 kg
YieldFor 8.2 L medium
Product TypeDehydrated Culture Media
Unit SizeEach