Oxoid™ Bax™ E.Coli 0157:H7 MP Medium
Thermo Scientific™
Oxoid™ Bax™ E.Coli 0157:H7 MP Medium
The Bax™ E.Coli 0157:H7 MP medium from Oxoid is a selective agar for the isolation and differentiation of enterohemorrhagic (EHEC) E. coli O157:H7 strains from food and clinical material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM1076R2.5 kg
CM1076G10 kg
Catalog number CM1076R
Price (INR)
-
Quantity:
2.5 kg
The Bax™ E.Coli 0157:H7 MP medium from Oxoid is a selective agar for the isolation and differentiation of enterohemorrhagic (EHEC) E. coli O157:H7 strains from food and clinical material.
Specifications
For Use With (Application)For the isolation and differentiation of enterohemorrhagic E. coli O157:H7 strains from food and clinical material
Quantity2.5 kg
Product TypeMicrobiology Media
Unit SizeEach