Oxoid™ Cronobacter Screening Broth Base
Oxoid™ Cronobacter Screening Broth Base
Oxoid™ Cronobacter Screening Broth Base
Thermo Scientific™
Oxoid™ Cronobacter Screening Broth Base
Oxoid Cronobacter Screening Broth Base, a selective enrichment broth, isolates Cronobacter spp, from food and environmental samples.
Have Questions?
Catalog NumberQuantity
CM1121B500 g
Catalog number CM1121B
Price (INR)
-
Quantity:
500 g
Oxoid Cronobacter Screening Broth Base (CSB) is a selective enrichment broth for isolation of Cronobacter spp, from food and environmental samples.
Requires Vancomycin Supplement, 5mg, Part No. SR0247E.
For Laboratory Use Only
Specifications
DescriptionCronobacter Screening Broth Base
Quantity500 g
YieldFor 17.8L medium
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach