Product Image
Gibco™
Cryopreserved Hepatocyte Plating Medium (CHPM™)
Cryopreserved Hepatocyte Plating Medium (CHPM®) used for plating Gibco cryopreserved human hepatocytes for optimal attachment and monolayer formation.Related linkLearn moreRead more
Have Questions?
Catalog NumberQuantity
CM900050 mL
Catalog number CM9000
Price (INR)
-
Quantity:
50 mL
Cryopreserved Hepatocyte Plating Medium (CHPM®) used for plating Gibco cryopreserved human hepatocytes for optimal attachment and monolayer formation.
Related link
Learn more about our Hepatic Cell Culture reagents
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHepatocyte Cells
Product LineGibco
Quantity50 mL
RemarksProprietary formulation provided by AP Sciences
Shipping ConditionWet Ice
ClassificationFBS-supplemented
FormLiquid
Product TypeHepatocyte Supplement
SterilitySterile
With AdditivesPhenol Red
Unit SizeEach
Contents & Storage
Store at 4°C