Sensititre™ NARMS Gram Positive CMV3AGPF AST Plate
Sensititre™ NARMS Gram Positive CMV3AGPF AST Plate
Sensititre™ NARMS Gram Positive CMV3AGPF AST Plate
Thermo Scientific™
Sensititre™ NARMS Gram Positive CMV3AGPF AST Plate
Comply with US National Antimicrobial Resistance Monitoring System (NARMS) surveillance mandates for Enterococci with this convenient plate.
Have Questions?
Catalog NumberQuantity
CMV3AGPF10/Pk.
Catalog number CMV3AGPF
Price (INR)
-
Quantity:
10/Pk.
Test Enterococcus isolates as a part of the National Antimicrobial Resistance Monitoring System (NARMS) program in the United States with Sensititre™ NARMS Gram Positive CMV3AGPF AST Plate.
Comply with US National Antimicrobial Resistance Monitoring System (NARMS) surveillance mandates for Enterococci with Sensititre NARMS Gram Positive CMV3AGPF AST Plate.
  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows the laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Automated or manual read options are available for ultimate flexibility.
The National Antimicrobial Resistance Monitoring System (NARMS) is a U. S. public health surveillance system that tracks antimicrobial resistance in foodborne and other enteric bacteria. The Sensititre system is an end-to-end solution for susceptibility testing that meets the requirements of the National Antimicrobial Resistance Monitoring System (NARMS).
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionSensititre NARMS Gram Positive Plate
Isolates Per Plate1
Quantity10/Pk.
Reading OptionsAutoread or manual
Product TypeAST Plate
SpeciesEnterococcus species
Target Organism ClassGram-positive Organisms
Test TypeAntimicrobial Surveillance
Unit SizeEach