Sensititre™ NARMS Gram Negative CMV4AGNF AST Plate
Thermo Scientific™
Sensititre™ NARMS Gram Negative CMV4AGNF AST Plate
Comply with US National Antimicrobial Resistance Monitoring System (NARMS) surveillance mandates for Salmonella and Escherichia coli with this plate.
Have Questions?
Catalog NumberQuantity
CMV4AGNF10 Plates/Box
Catalog number CMV4AGNF
Price (INR)
-
Quantity:
10 Plates/Box
Perform antimicrobial susceptibility testing for Salmonella and Escherichia coli isolates as a part of the National Antimicrobial Resistance Monitoring System (NARMS) program in the United States with Sensititre™ NARMS Gram Negative CMV4AGNF AST Plate.
Comply with US National Antimicrobial Resistance Monitoring System (NARMS) surveillance mandates for Salmonella and Escherichia coli with Sensititre NARMS Gram Negative CMV4AGNF AST Plate.
  • Extended shelf life, room temperature storage eliminates inventory control concerns
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Automated or manual read options are available for ultimate flexibility
The National Antimicrobial Resistance Monitoring System (NARMS) is a U. S. public health surveillance system that tracks antimicrobial resistance in foodborne and other enteric bacteria. The Sensititre system is an end-to-end solution for susceptibility testing that meets the requirements of the National Antimicrobial Resistance Monitoring System (NARMS).
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionSensititre NARMS Gram Negative CMV4AGNF AST Plate
Isolates Per Plate1
No. per Pack10
Packaging TypeBox
Quantity10 Plates/Box
Reading OptionsManual, Vizion, OptiRead, ARIS HiQ
Regulatory StatusRUO
Product TypeAST Plate
SpeciesSalmonella species, Escherichia coli
Target Organism ClassGram-negative Microorganisms
Test TypeAntimicrobial Surveillance
Unit SizeEach