LN<sub>2</sub> Level Monitor
Actual product may vary
Thermo Scientific™
LN2 Level Monitor
Safeguard irreplaceable biological specimens with the LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Voltage
CN509X4-70Locator 4 Plus100/240 V
CN509X1Locator Jr.100/240 V
CN509X1-70Locator Jr. Plus100/240 V
CN509X4Locator 4100/240 V
CN509X11-70Locator 6 Plus100/240 V
CN509X7Locator 4 Plus230 V
Catalog number CN509X4-70
Price (INR)
-
For Use With (Equipment):
Locator 4 Plus
Voltage:
100/240 V
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
  • Minimizes conduction and liquid nitrogen evaporation
  • Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
  • Audible and visual alarms signal low level liquid nitrogen conditions
  • Warranty varies by country, please contact us for details
Specifications
DescriptionLevel Monitor
For Use With (Equipment)Locator 4 Plus
TypeLN2 Measuring Device
Monitoring OptionsAlarm
Voltage100/240 V
Unit SizeEach