Sensititre™ Companion Animal MIC Plate
Actual product may vary
Sensititre™ Companion Animal MIC Plate
Sensititre™ Companion Animal MIC Plate
Thermo Scientific™
Sensititre™ Companion Animal MIC Plate
Perform accurate AST with companion animal-specific antimicrobics, including cefovecin, with the Thermo Scientific™ Sensititre™ Companion Animal MIC Plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
COMPAN2FMIC Plate with Cephalothin
COMPAN1FMIC Plate
Catalog number COMPAN2F
Price (INR)
-
Description:
MIC Plate with Cephalothin
Perform accurate AST with companion animal-specific antimicrobics, including cefovecin, with the Thermo Scientific™ Sensititre™ Companion Animal MIC Plate.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionMIC Plate with Cephalothin
Isolates Per Plate1
No. per Pack10
Quantity10/Pk.
Reading OptionsAutoread or manual
Product TypeMIC Plate
SpeciesCompanion Animals
Target Organism ClassGram-negative Organisms, Gram-positive Organisms
Test TypeMinimum Inhibitory Concentration (MIC)
Unit SizeEach