Oxoid™ Mupirocin Antimicrobial Susceptibility discs
Oxoid™ Mupirocin Antimicrobial Susceptibility discs
Oxoid™ Mupirocin Antimicrobial Susceptibility discs
Thermo Scientific™
Oxoid™ Mupirocin Antimicrobial Susceptibility discs
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
CT0523B200 μg
CT1826B20 μg
Catalog number CT0523B
Price (INR)
-
Concentration:
200 μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Antibiotic CodeMUP
Antimicrobial AgentsMupirocin
Certifications/ComplianceComplies with EUCAST guidelines
Concentration200 μg
DescriptionOxoid Mupirocin Antimicrobial Susceptibility Discs
No. per Pack5 Cartridges/Pk.
Packaging Type50 Discs/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit SizeEach