Oxoid™ Apramycin Antimicrobial Susceptibility Disks, 15 μg
Oxoid™ Apramycin Antimicrobial Susceptibility Disks, 15 μg
Oxoid™ Apramycin Antimicrobial Susceptibility Disks, 15 μg
Thermo Scientific™
Oxoid™ Apramycin Antimicrobial Susceptibility Disks, 15 μg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Have Questions?
Catalog NumberConcentration
CT0545B15 μg
Catalog number CT0545B
Price (INR)
-
Concentration:
15 μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Apramycin Antimicrobial Susceptibility Disks.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For Veterinary Use Only.
For Veterinary Use Only.
Specifications
Antibiotic CodeAPR
Antimicrobial AgentsApramycin
Certifications/ComplianceComplies with CLSI guidelines
Concentration15 μg
DescriptionOxoid Apramycin Antimicrobial Susceptibility Disks
No. per Pack5 Cartridges/Pk.
Packaging Type50 Disks/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit SizeEach