Oxoid™ Aztreonam-Avibactam Antimicrobial Susceptibility Discs
Thermo Scientific™

Oxoid™ Aztreonam-Avibactam Antimicrobial Susceptibility Discs

Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing discs.
Have Questions?
Catalog NumberQuantity
CT1971B50 Discs/Cartridge
Catalog number CT1971B
Price (INR)
-
Quantity:
50 Discs/Cartridge
Manually determine the antibiotic susceptibility of microorganisms using disc diffusion methods with consistently reliable, industry-leading quality1 Thermo Scientific™ Oxoid™ Aztreonam-Avibactam Antimicrobial Susceptibility Discs.

The drug aztreonam-avibactam has been designed to address infections caused by aerobic Gram-negative organisms that are resistant to other antibiotics due to the production of beta-lactamase enzymes. Aztreonam-avibactam is especially valuable for infections caused by organisms producing metallo-beta-lactamases (MBLs), which are resistant to many other beta-lactam antibiotics.

Perform susceptibility testing of multi-drug resistant aerobic Gram-negative organisms using Aztreonam-Avibactam Antimicrobial Susceptibility Discs. 

Features include:

  • Meets EUCAST and CLSI recommended breakpoints
  • Produces quality results in line with the rest of the Oxoid disc portfolio as demonstrated over 3 EUCAST disc quality studies
  • Easy-to-use: shatterproof spring-loaded cartridges, easy-to-read labelling, and end-of-cartridge indicators
  • Long-term stability ensured: each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%
  • Accurate and reproducible: manufactured under stringent quality standards including BS EN ISO 9001:2008, ISO 13485:2003, and FDA 21 CFR part 820, with comprehensive quality control testing
  • Trusted supplier: over 35 years of experience in disc development and manufacture

Aztreonam-Avibactam has been shown to be effective against:

  • Escherichia coli
  • Klebsiella pneumoniae
  • Klebsiella oxytoca
  • Enterobacter cloacae complex
  • Citrobacter freundii complex
  • Serratia marcescens

Disc diffusion not only categorizes resistant, intermediate, and sensitive organisms through qualitative results but also provides a visual indication of the following:

  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics

Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.

For In Vitro Diagnostic Use.  
  1. Åhman J, Matuschek E, Kahlmeter G. The quality of antimicrobial discs from nine manufacturers—EUCAST evaluations in 2014 and 2017. Clin Microbiol Infect. 2019; 25: 346-352. https://www.clinicalmicrobiologyandinfection.com/action/showPdf?pii=S1198-743X%2818%2930456-7
Specifications
Antibiotic CodeAZA
Antimicrobial AgentsAztreonam-Avibactam
Certifications/ComplianceComplies with CLSI & EUCAST guidelines
Concentration30/20µg
No. per Pack5 Cartridges/Pk 
Packaging Type250 Discs/Box
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Target Organism ClassGram-negative Microorganisms
Test TypeAntimicrobial Susceptibility for Gram-negative Organisms
Unit SizeEach