Product Image
Thermo Scientific™
Remote Controller Cable, which connects Neslab units to a computer, TC-400 Remote Box, or RPC Touchscreen Programmable Controller
These Remote Controller Cables enable communication from a Thermo Scientific water bath to a computer.
Have Questions?
Catalog NumberType
CTD-16032Remote Controller Cables
Catalog number CTD-16032
Price (INR)
-
Type:
Remote Controller Cables
Thermo Scientific™ Remote Controller Cables can be used to connect a Thermo Scientific bath equipped with an RS-232 or RS-485 interface port to a computer, TC-400 Remote Box or the new RPC Touchscreen Programmable Controller.
Available Cable Lengths:
  • 10ft. (3.0m)
  • 25ft. (7.6m)
  • 50ft. (15.2m)
  • 75ft. (22.9m)
  • 100ft. (30.5m)
  • 200ft. (61.0m)
Specifications
CommunicationsRS232, RS485
For Use With (Equipment)Connects Neslab units to a computer, TC-400 Remote Box, or RPC Touchscreen Programmable Controller
TypeRemote Controller Cables
Unit SizeEach