Specifications
Activity2 ng/ml
BioassayProliferation of MO7e cells
ClassificationCarrier-Free
Endotoxin Level<0.1 ng/μg
Expression SystemE. coli
Gene AliasSCF (c-kit Ligand)
Molecular Weight (g/mol)18.5 kDa
Product LineGibco
Protein FamilyGrowth Factors & Receptors
Protein FormFull Length, Recombinant, Ligand
Protein Length164 aa
Protein SubtypeOther Proteins
Purification MethodSequential Chromatography
Purity or Quality Grade95 %
Quantity1 mg
Research CategoryClinical Research, Stem Cell Research, Differentiation, Oncology
Shipping ConditionWet Ice
SourceHuman
For Use With (Application)Cell Culture
FormLyophilized
RecombinantRecombinant
SpeciesHuman
Unit Size1 mg