Benefit from automatic operation with these polythylene and fiberglass storage reservoirs for Thermo Scientific™ Barnstead™ reverse osmosis water systems.
Have Questions?
Change view
Catalog Number
Capacity (English)
D12681
8 gal.
B5041
50 gal.
B5042
100 gal.
B5043
200 gal.
D9021
16 gal.
5 Options
Catalog number D12681
Price (INR)
-
Capacity (English):
8 gal.
Benefit from automatic operation with polythylene and fiberglass Thermo Scientific™ Barnstead™ RO Storage Reservoirs. For use with Barnstead RO (Reverse Osmosis) water systems.
Polyethylene Reservoirs:
Control panel that displays reservoir's water level
Vent protected to eliminate the introduction of airborne contamination
Draw-off points that include spigot, dishwasher port (for 60 and 100L reservoirs only), and feed to Thermo Scientific™ Nanopure™ systems
Can be wall- or bench-mounted
Constructed of rigid fluorinated polyethylene, providing strength and protection against material contamination of the water
Pump protector provided; ensures the system is not damaged when there is not enough water in the reservoir
Single Access Port:
Single port to connect reservoir to other systems and/or equipment
Capacity: 30L
Multiple Access Port:
Optional distribution pump available
Perfect for supplying water to a remote Nanopure or glassware washer: easily mounts to base of 16 gal. (60L) reservoir and can supply reverse osmosis water up to 35′ (10.7m) away
Capable of delivering up to 1 gal. (3.8L) of water/min. at 15psig (1.3 bar)
Automatically circulates water for 15 minutes every hour when in standby
Multiple ports to connect reservoir to other systems and/or equipment