Dextran, Tetramethylrhodamine, 10,000 MW, Anionic, Fixable
Invitrogen™

Dextran, Tetramethylrhodamine, 10,000 MW, Anionic, Fixable

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D1868
also known as D-1868
25 mg
Catalog number D1868
also known as D-1868
Price (INR)
16,662.00
Online offer
Ends: 31-Dec-2026
24,684.00
Save 8,022.00 (32%)
Each
Quantity:
25 mg
Price (INR)
16,662.00
Online offer
Ends: 31-Dec-2026
24,684.00
Save 8,022.00 (32%)
Each
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

• Label (Ex/Em): Tetramethylrhodamine (555/580)
• Size: 10,000 MW
• Charge: Anionic
• Fixable: Fixable via free amines

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor™, Cascade Blue™, lucifer yellow, fluorescein, and Oregon Green™ dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor™ dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

• Neuronal tracing (anterograde and retrograde) in live cells
• Cell lineage tracing in live cells
• Neuroanatomical tracing
• Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
• Investigating vascular permeability and blood–brain barrier integrity
• Tracking endocytosis
• Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
• Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeClassic Dyes
Product TypeDextran
Quantity25 mg
Shipping ConditionRoom Temperature
Excitation/Emission555/580 nm
Product LineInvitrogen
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (15)

Citations & References
Abstract
Analysis of the growth cone turning assay for studying axon guidance.
Authors:Pujic Z, Giacomantonio CE, Unni D, Rosoff WJ, Goodhill GJ,
Journal:J Neurosci Methods
PubMed ID:18313760
The
Diffusion of microinjected markers across the parasitophorous vacuole membrane in cells infected with Eimeria nieschulzi (Coccidia, Apicomplexa).
Authors:Werner-Meier R, Entzeroth R
Journal:Parasitol Res
PubMed ID:9211514
Cells infected with the intracellular parasite Eimeria nieschulzi were microinjected with lucifer yellow (457 Da), biocytin lucifer yellow (850 Da) and dextranrhodamine (10,000 Da). Immediately after injection of a mixture of the markers into the host cell cytoplasm, a differential diffusion pattern was observed in trophozoites and schizonts. Lucifer yellow ... More
Monitoring autophagy in lysosomal storage disorders.
Authors:Raben N, Shea L, Hill V, Plotz P,
Journal:Methods Enzymol
PubMed ID:19216919
Lysosomes are the final destination of the autophagic pathway. It is in the acidic milieu of the lysosomes that autophagic cargo is metabolized and recycled. One would expect that diseases with primary lysosomal defects would be among the first systems in which autophagy would be studied. In reality, this is ... More
Neural organization and visual processing in the anterior optic tubercle of the honeybee brain.
Authors:Mota T, Yamagata N, Giurfa M, Gronenberg W, Sandoz JC,
Journal:J Neurosci
PubMed ID:21832175
The honeybee Apis mellifera represents a valuable model for studying the neural segregation and integration of visual information. Vision in honeybees has been extensively studied at the behavioral level and, to a lesser degree, at the physiological level using intracellular electrophysiological recordings of single neurons. However, our knowledge of visual ... More
Persistence of gap junction communication during myocardial ischemia.
Authors:Ruiz-Meana M, Garcia-Dorado D, Lane S, Pina P, Inserte J, Mirabet M, Soler-Soler J
Journal:Am J Physiol Heart Circ Physiol
PubMed ID:11356611
During myocardial ischemia, severe ATP depletion induces rigor contracture followed by intracellular Ca2+ concentration ([Ca2+]i) rise and progressive impairment of gap junction (GJ)-mediated electrical coupling. Our objective was to investigate whether chemical coupling through GJ allows propagation of rigor in cardiomyocytes and whether it persists after rigor development. In end-to-end ... More