Barnstead™ Easypure™ RoDi Accessories and Consumables
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Have Questions?
Change view
Catalog Number
Description
D502132
Start-up Kit
AY1332X1
External Booster Pump
AY1332X2
Pre-filtration Accessory Kit
CV742X5A
Ventgard Reservoir Cap
D13324
Wall Mounting Bracket
D7034
Empty Cartridges
FL1332X1
Reverse Osmosis Membrane
7 Options
Catalog number D502132
Price (INR)
-
Description:
Start-up Kit
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.