Dextran, Oregon Green™ 488; 10,000 MW, Anionic
Invitrogen™

Dextran, Oregon Green™ 488; 10,000 MW, Anionic

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D71705 mg
Catalog number D7170
Price (INR)
25,713.00
キャンペーン価格
Ends: 31-Dec-2026
42,855.00
Save 17,142.00 (40%)
Each
Quantity:
5 mg
Price (INR)
25,713.00
キャンペーン価格
Ends: 31-Dec-2026
42,855.00
Save 17,142.00 (40%)
Each
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

• Label (Ex/Em): Oregon Green™ 488 (496/524)
• Size: 10,000 MW
• Charge: Anionic
• Fixable: Nonfixable

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor™ 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor™, Cascade Blue™, lucifer yellow, fluorescein, and Oregon Green™ dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor™ dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

• Neuronal tracing (anterograde and retrograde) in live cells
• Cell lineage tracing in live cells
• Neuroanatomical tracing
• Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
• Investigating vascular permeability and blood–brain barrier integrity
• Tracking endocytosis
• Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
• Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeOther Label(s) or Dye(s)
Product TypeDextran
Quantity5 mg
Shipping ConditionRoom Temperature
Excitation/Emission496/524 nm
Product LineOregon Green
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (26)

Citations & References
Abstract
The CRE/CREB pathway is transiently expressed in thalamic circuit development and contributes to refinement of retinogeniculate axons.
Authors:Pham TA, Rubenstein JL, Silva AJ, Storm DR, Stryker MP
Journal:Neuron
PubMed ID:11516398
'The development of precise connections in the mammalian brain proceeds through refinement of initially diffuse patterns, a process that occurs largely within critical developmental windows. To elucidate the molecular pathways that orchestrate these early periods of circuit remodeling, we have examined the role of a calcium- and cAMP-regulated transcriptional pathway. ... More
Development of a delivery vehicle for intracellular transport of botulinum neurotoxin antagonists.
Authors:Goodnough MC, Oyler G, Fishman PS, Johnson EA, Neale EA, Keller JE, Tepp WH, Clark M, Hartz S, Adler M
Journal:FEBS Lett
PubMed ID:11904143
'A targeted delivery vehicle (DV) was developed for intracellular transport of emerging botulinum neurotoxin (BoNT) antagonists. The DV consisted of the isolated heavy chain (HC) of BoNT/A coupled to a 10-kDa amino dextran via the heterobifunctional linker 3-(2-pyridylthio)-propionyl hydrazide. The HC served to target BoNT-sensitive cells and promote internalization of ... More
Loss of the chloride channel ClC-7 leads to lysosomal storage disease and neurodegeneration.
Authors:Kasper D, Planells-Cases R, Fuhrmann JC, Scheel O, Zeitz O, Ruether K, Schmitt A, Poët M, Steinfeld R, Schweizer M, Kornak U, Jentsch TJ
Journal:EMBO J
PubMed ID:15706348
'ClC-7 is a chloride channel of late endosomes and lysosomes. In osteoclasts, it may cooperate with H(+)-ATPases in acidifying the resorption lacuna. In mice and man, loss of ClC-7 or the H(+)-ATPase a3 subunit causes osteopetrosis, a disease characterized by defective bone resorption. We show that ClC-7 knockout mice additionally ... More
Dermal microvascular endothelial cells express the 180-kDa macrophage mannose receptor in situ and in vitro.
Authors:Gröger M, Holnthoner W, Maurer D, Lechleitner S, Wolff K, Mayr BB, Lubitz W, Petzelbauer P
Journal:J Immunol
PubMed ID:11067894
'Expression of the 180-kDa mannose receptor (MR) is mainly found on cells of the macrophage lineage. MR mediates the uptake of micro-organisms and host-derived glycoproteins. We demonstrate that endothelium of the human skin in situ and dermal microvascular endothelial cells (DMEC) in vitro expressed MR at both the protein and ... More
Characterization of cellular optoporation with distance.
Authors:Soughayer JS, Krasieva T, Jacobson SC, Ramsey JM, Tromberg BJ, Allbritton NL
Journal:Anal Chem
PubMed ID:10740880
'We have developed and characterized cellular optoporation with visible wavelengths of light using standard uncoated glass cover slips as the absorptive media. A frequency-doubled Nd:YAG laser pulse was focused at the interface of the glass surface and aqueous buffer, creating a stress wave and transiently permeabilizing nearby cells. Following optoporation ... More