Product Image
Thermo Scientific™

Gene Pool™ cDNA, from human adult normal vagina tissue

In addition to the previously listed Gene Pool™ cDNA, the following selection of cDNA from adult human normal, tumor, orRead more
Have Questions?
Catalog NumberQuantity
D814001
also known as D8140-01
20 Reactions
Catalog number D814001
also known as D8140-01
Price (INR)
-
Quantity:
20 Reactions
In addition to the previously listed Gene Pool™ cDNA, the following selection of cDNA from adult human normal, tumor, or disease tissues as well as human fetal normal tissue is available. See Gene Pool™ cDNA for description and storage information. All Special Order Products are subject to tissue/tumor availability. Please call for information. Allow up to two weeks for delivery.
Specifications
No. of Reactions20 reactions
Product LineGene Pool
Product TypecDNA
Quantity20 Reactions
Sample SourceNormal Tissue
Unit Size20 reactions