Barnstead™ Hose Nipple Cartridges
Thermo Scientific™

Barnstead™ Hose Nipple Cartridges

Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionResin Type
D89512/3 Size Mixed-Bed and Organic RemovalMixed-bed Ion Exchange with Activated Carbon
D502201/2 Size Mixed-BedUltrapure Ion Exchange
D88221/2 Size Mixed-Bed with Oxygen RemovalUltrapure Mixed Bed with Strong Anion Exchange
D8901Full Size High CapacityTwo Bed Ion Exchange
D8902Full Size UltrapureUltrapure Mixed Bed Ion Exchange
D8903Full Size Oxygen RemovalStrong Anion Exchange
D8904Full Size Organic RemovalActivated Carbon
D8905Hose nipple cartridge, Full Size Cation RemovalCation Exchange
D8908Full Size Macroreticular, Cation RemovalMacroreticular Cation Exchange
D8911Full Size UltrapureUltrapure Ion Exchange
D8921Full Size Pretreatment and Scale EliminatorMixed-bed and Strong Cation Exchange, Activated Charcoal
D8922Full Size Ultrapure with Organic RemovalMixed-bed Ion Exchange with Activated Carbon
D89502/3 Size High CapacityTwo Bed Ion Exchange with Strong Cation Exchange
D8900Hose Nipple Cartridge Holder
Catalog number D8951
Price (INR)
-
Description:
2/3 Size Mixed-Bed and Organic Removal
Resin Type:
Mixed-bed Ion Exchange with Activated Carbon

Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.

  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8 in. barb hose connections on each end
  • D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8 in. ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
Specifications
Description2/3 Size Mixed-Bed and Organic Removal
Flow RateUp to 75 L/hr.
For Use With (Application)Mixed-bed Ion Exchange with Activated Carbon
IncludesColor Indicator
Resin TypeMixed-bed Ion Exchange with Activated Carbon
For Use With (Equipment)Barnstead Water Purification System
Particle Retention470 grains/2000 gal.
TypeCartridge, Hose Nipple
Unit SizeEach