Dry-Bags™ Maximum Recovery Diluent (MRD)
Thermo Scientific™

Dry-Bags™ Maximum Recovery Diluent (MRD)

Simplify media preparation in food laboratories with this high quality dehydrated medium in a convienient bag format; just open-fill-dispense, saving precious time in the lab.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatYield
DB0733WDry-Bags, without filter20L
DB0733MDry-Bags™, with filter20L
Catalog number DB0733W
Price (INR)
-
Format:
Dry-Bags, without filter
Yield:
20L
Save time in preparing bulk media and diluents for food microbiology testing using Thermo Scientific™ Dry-Bags™ Maximum Recovery Diluent (MRD). The irradiated media is supplied in lightweight, transparent plastic bags — just add water and the medium is ready for use. Each bag makes 20L of medium, which can be dispensed through the outlet tube.

Use Maximum Recovery Diluent (MRD) as a medium which combines the protective effect of peptone in the diluting solution with the osmotic support of physiological saline. The low concentration of peptone does not cause multiplication of the organisms within 45 minutes (@ 20-25°C) of dilution of the sample. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions.

Convenient

  • Quick and easy to prepare — just add deionized water through filter sterilization to each Dry-Bag, and they are ready to use in minutes. Pre-sterilized water can also be added under aseptic conditions.

Cost-Effective

  • Light to transport — there is no weighing of media or sterilization required, reducing the burden on media preparation departments and equipment.

Safe

  • No handling of glassware or weighing of culture media required, no need to add supplementary reagents — empty bags can be disposed of with normal laboratory waste.

Quality Guaranteed

  • Quality certificates are available for every batch to help reduce the burden of QC testing.

Workflow

  • Dry-Bags are easily adaptable to laboratory workflows and equipment such as automated diluters.

Easy-To-Use

  • Dry-Bags are easy to fill and dispense with minimum training.

Space-Saving

  • Conveniently packed in flat boxes of five or ten, Dry-Bags take up relatively little space compared to prepared media and can be made up as required.

Re-hydrated Dry-Bags medium can be stored for 3 days at room temp (away from strong light sources) and are available with filters attached for convenience or without filters as a lower-cost option. For those Dry-Bags supplied without filters, autoclavable, reusable filters are also available (DB0200A). For added convenience, it is recommended that the Dry-Bags be filled using the Thermo Scientific Peristaltic Pump (DB2000A or DB3000A) and dispensed using the Thermo Scientific Automated Diluter (DB4000A).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
ColorClear, colorless
DescriptionDry-Bags Maximum Recovery Diluent (MRD)
FormatDry-Bags, without filter
Quantity10/Pk.
Shelf Life2 Years
Yield20L
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
10°C to 30°C