Product Image
Thermo Scientific™
Oxoid™ X Factor Discs
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging Type
DD0003T1 Cart./Pk.
DD0003B5 Cart./Pk.
Catalog number DD0003T
Price (INR)
-
Packaging Type:
1 Cart./Pk.
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Ordering Alerts:
50 Discs/Cartridge
Specifications
DescriptionX Factor Discs
Packaging Type1 Cart./Pk.
Quantity50 Disks/Cartridge
TypeX Factor Disc
Detectable AnalytesHaemophilus species
Unit SizeEach
Contents & Storage
-10°C to -20°C