See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Microbial Identification Test Kits
▸
Haemophilis Testing
▸
Oxoid™ X + V Factor Disc
Thermo Scientific™
Oxoid™ X + V Factor Disc
Oxoid X + V Factor Disc is used for identification of
Haemophilus
spp.
Have Questions?
Catalog Number
Detectable Analytes
DD0005T
Haemophilus species
Catalog number
DD0005T
Price (INR)
-
Request A Quote
Detectable Analytes:
Haemophilus species
Oxoid X + V Factor Disc is used for identification of
Haemophilus
spp.
50 Discs/Cartridge
Specifications
Description
X + V Factor Disk
Packaging Type
1 Cart./Pk.
Type
V Factor Disc
Detectable Analytes
Haemophilus species
Unit Size
Each
Contents & Storage
-10°C to -20°C