An-Ident Discs
Thermo Scientific™
An-Ident Discs
Presumptively identify anaerobic Gram-negative bacteria using these diagnostic discs.
Have Questions?
Catalog NumberQuantity
DD0006A50 Discs/Cartridge
Catalog number DD0006A
Price (INR)
-
Quantity:
50 Discs/Cartridge
Presumptively identify anaerobic Gram-negative bacteria using Thermo Scientific™ Oxoid™ An-Ident Discs. Based on the differences in response to a number of antibiotics, Sutter and Finegold1 proposed a method for identification of Gram-negative anaerobic bacilli. Using carefully chosen levels of colistin, erythromycin, kanamycin, vancomycin penicillin and rifampicin with disc diffusion sensitivity testing techniques, they showed that these bacteria could be placed fairly consistently into five groups. Other simple biochemical tests and cultural characteristics were used for further identification.
Use An-Ident Discs as a rapid and simple method for presumptive identification of anaerobic Gram-negative bacteria. They are primarily designed to allow easy identification of Bacteroids fragilis, the anaerobe species most commonly isolated in the routine hospital laboratory.
Rapid Results
  • Simple and ready-to-use in cartridge format
  • Suitable for use either in the Thermo Scientific Oxoid Dispenser or with Thermo Scientific Oxoid Ejectors
Antibiotic levels on the discs are as follows:
Erythromycin (code E60) - 60μg
Rifampicin (code RD15) - 15μg
Colistin (code CT10) - 10μg
Penicillin (code P2) - 2 units
Kanamycin (code K1000) - 1000μg
Vancomycin (code VA5) - 5μg
An-Ident Discs are not suitable for antibiotic susceptibility testing to assess the clinical effectiveness of the various antibiotics.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
General References:
  1. Sutter, V.L. and Finegold, S.M. (1971) Antibiotic Disc Susceptibility Tests for Rapid Presumptive Identification of Gram-negative Anaerobic Bacilli. Appl. Microbiol.. 21, 13 - 20.
  2. Leigh, D.A. and Simmons, K. (1977) Identification of non-sporing anaerobic bacteria J. Clin. Path 30, 991-992.
Specifications
DescriptionAn-Ident Discs
FormatDisk, Cartridge
Packaging Type6 Cart./Pk.
Quantity50 Discs/Cartridge
TypeAn-Ident Disk
Detectable AnalytesAnaerobic Gram-negative Bacteria
Unit SizeEach
Contents & Storage
-20°C to +8°C