Product Image
Thermo Scientific™
Oxoid™ Metronidazole Disc
Thermo Scientific™ Oxoid Metronidazole Disc is used for presumptive identification of Gardnerella vaginalis.
Have Questions?
Catalog NumberQuantity
DD0008T50 Disks
Catalog number DD0008T
Price (INR)
-
Quantity:
50 Disks
Thermo Scientific™ Oxoid Metronidazole Disc is used for presumptive identification of Gardnerella vaginalis.
Specifications
DescriptionMetronidazole Disk
FormatDisk, Cartridge
Packaging Type1 Cart./Pk.
Quantity50 Disks
TypeMetronidazole Disk
Detectable AnalytesG. vaginalis
Unit SizeEach
Contents & Storage
-20°C