Product Image
Thermo Scientific™
Oxoid™ Sulphonamide Disc
Thermo Scientific™ Oxoid Sulphonamide Disc is used for identification of Gardnerella vaginalis.
Have Questions?
Catalog NumberQuantity
DD0011T50 Disks/Cartridge
Catalog number DD0011T
Price (INR)
-
Quantity:
50 Disks/Cartridge
Thermo Scientific™ Oxoid Sulphonamide Disc is used for identification of Gardnerella vaginalis.
  • 100μg
  • 50 Discs/Cartridge
Specifications
DescriptionSulphonamide Disk
FormatDisk, Cartridge
Packaging Type1 Cart./Pk.
Quantity50 Disks/Cartridge
TypeSulphonamide Disk
Detectable AnalytesG. vaginalis
Unit SizeEach
Contents & Storage
2°C to 8°C