Product Image
Thermo Scientific™
Oxoid™ SPS Disc
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Have Questions?
Catalog NumberQuantity
DD0016T50 Disks/Cartridge
Catalog number DD0016T
Price (INR)
-
Quantity:
50 Disks/Cartridge
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Susceptibility disc format
Specifications
DescriptionSPS Disk
FormatSusceptibility Disk
Quantity50 Disks/Cartridge
TypeSPS Disk
Detectable AnalytesP. anaerobius
Unit SizeEach