Kanamycin 1000μg Discs
Thermo Scientific™
Kanamycin 1000μg Discs
Presumptively identify Gram-negative anaerobic bacilli using these diagnostic discs.
Have Questions?
Catalog NumberQuantity
DD0027B50 Discs/Cartridge (250 Discs/Pk.)
Catalog number DD0027B
Price (INR)
-
Quantity:
50 Discs/Cartridge (250 Discs/Pk.)
Presumptively identify Gram-negative anaerobic bacilli with Thermo Scientific™ Oxoid™ Kanamycin 1000μg Discs. Kanamycin is an aminoglycoside which, in high concentrations, can be used in combination with routine susceptibility discs to presumptively identify Gram-negative, anaerobic bacilli.
Use Kanamycin 1000μg Discs as a rapid and simple method for presumptive identification of anaerobic Gram-negative bacteria.
  • Rapid results
  • Simple and ready-to-use cartridge format.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration1000 μg/disc
DescriptionKanamycin Discs
FormatDisk, Cartridge
Packaging Type5 Cart./Pk.
Quantity50 Discs/Cartridge (250 Discs/Pk.)
TypeKanamycin Disk
Detectable AnalytesAnaerobic Gram-negative Bacteria
Unit SizeEach
Contents & Storage
-20°C to +8°C