Dog (Beagle) Microsomes
Product Image
Gibco™

Dog (Beagle) Microsomes

Liver microsomes prepared from a pool of male Beagle Dogs.Total protein concentration: 20 mg/mlWe provide liver subcellular fractions from aRead more
Have Questions?
Catalog NumberQuantity
DGMCPL0.5 mL
Catalog number DGMCPL
Price (INR)
11,027.00
Each
Add to cart
Quantity:
0.5 mL
Price (INR)
11,027.00
Each
Add to cart
Liver microsomes prepared from a pool of male Beagle Dogs.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20 mg/mL
Donor SourcePooled
GenderMale
Quantity0.5 mL
Shipping ConditionDry Ice
Sub Cellular LocalizationMicrosomes
Target SpeciesDog/Canine
TissueLiver
FormFrozen
Product LineGibco
Product TypeSubcellular Fractions
Unit SizeEach
Contents & Storage
Store between -60°C and -85°C