Thermo Scientific™ Oxoid DrySpot test is a screen for culture confirmation of E. coli serogroups O26, O91, O103, O111, O128 and O145.
Have Questions?
Catalog Number
Quantity
DR0300M
20 X 3 Tests/Kit
Catalog number DR0300M
Price (INR)
-
Quantity:
20 X 3 Tests/Kit
Thermo Scientific™ Oxoid DrySpot test is a latex agglutination screen used for culture confirmation of E. coli serogroups O26, O91, O103, O111, O128 and O145 from cultures on solid media.