Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles
Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles
Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles
Thermo Scientific™

Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles

Conduct light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth amber HDPE bottles.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
DS2085-000130 mL20 mm
Catalog number DS2085-0001
Price (INR)
7,575.00
Case of 72
Capacity (Metric):
30 mL
Closure Size:
20 mm
Price (INR)
7,575.00
Case of 72

Conduct light-duty, general purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Bottles. Lightweight versions of the original Nalgene Amber Lab Quality bottle, these high-density polyethylene bottles reduce UV light transmissions to protect light-sensitive liquids.

  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • Economical and recyclable; designed for single use
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

.

Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity1 oz., 30mL
Closure ColorAmber
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size20 mm
Closure TypeScrew Cap
DescriptionAmber Narrow-Mouth Economy Bottle
Diameter (Metric)34 mm
Diameter (Metric) Inner Neck14 mm
Height (English)2.4 in.
Height (Metric)61 mm
Neck Finish20mm
No. per Case72
No. per Pack72
Through the Closure DispensingNo
Width (English)1.35 in.
Capacity (English)1 oz.
Capacity (Metric)30 mL
Certifications/ComplianceMeets the requirements for light-resistant containers per USP latest edition
Closure IncludedYes
ColorOpaque Amber
DisposableSingle-use
GraduatedNo
MaterialHDPE
MouthNarrow
Product TypeNarrow Mouth Bottle
ShapeRound
SterilityNon-sterile
Unit SizeCase of 72