DataTaker DT80 Series Data Loggers
Thermo Fisher Scientific™
DataTaker DT80 Series Data Loggers
DataTaker is one of the world's leading brands of general purpose and specialised data loggers. DataTaker is supplier to broad range of customers across many industries including environmental, industrial, construction, manufacturing, process management, scientific, laboratory and education.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
DT80G-ALGeotechnical logger supporting vibrating wire strain gaugesLogger only (no accessories)
DT80General Purpose DataloggerLogger and accessories
DT80MGeneral Purpose DataloggerLogger w/ integrated modem and accessories
DT80WGeneral Purpose DataloggerLogger w/ integrated Wifi and accessories
DT80GGeotechnical logger supporting vibrating wire strain gaugesLogger and accessories
DT80-ALGeneral Purpose DataloggerLogger only (no accessories)
Catalog number DT80G-AL
Price (INR)
-
Type:
Geotechnical logger supporting vibrating wire strain gauges
Includes:
Logger only (no accessories)
DataTaker DT80 series comprises a family of data loggers, from five channel data loggers to systems with hundreds of analog inputs and many digital inputs. It measures and logs analog and digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, vibrating wire, strain gauges, current loops, voltages, resistance and frequencies. DataTaker models may include an Ethernet port, USB port, Modem for remote access and SMS alerts, WiFi, MODBUS connection, and may include a built-in web server, dEX, for setup and for viewing/downloading of data.
The DataTaker DT80 series comprises a family of industrial data loggers, ranging from five channel data loggers to systems with hundreds of analog inputs and many digital inputs.
The DataTaker DT80 series measures and logs analog & digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, strain gauges, current loops, voltages, resistance and frequencies.
DataTaker DT80 models include an Ethernet port, USB port and a built-in web server, dEX for setup and for viewing/downloading of data. In addition:
  • DT80M includes an integrated modem
  • DT80W and DT80GW include integrated WiFi
DataTaker DT80G is specifically suited to geotechnical applications such as slope stability, subsidence, dam wall monitoring, tunnel and mining excavation, ground water, tunnel wall monitoring and site assessment. DT80G supports vibrating wire sensors and other geotechnical sensors.
Other Features:
  • Analog input channels set up as binary logic level inputs.
  • Digital inputs recording ON/OFF states of contacts, or counting pulses over a desired period.
  • Alarm channels and digital outputs to raise alarms (email / SMS / physical digital outputs) or control external processes.
  • Readings may be returned directly or via internal calculations based on a single channel or multiple channels.
  • Internal memory holds ∼10 million readings.
  • Connection to the host computer via Ethernet, USB, and serial Host Port.
  • Separate USB port for USB flash drives to be used for extra storage, or automatic unloading of data from the logger.
  • Built-in battery or auxiliary battery option.
  • Most data logging configurations are easily set up using the graphical web-based dEX interface and a standard web browser.
  • For more complex configurations, a script can be developed using the logger's own scripting language.
Specifications
Battery TypeNone, external power required
CalibrationNot Calibrated
Channels5 to 15 Analog and 12 Digital (expandable using a CEM20 modules), high speed counter inputs and phase encoder inputs
Data Memory128 MB
Dimensions (L x W x H)180 x 137 x 65 mm
Humidity RangeMaximum: 85%, non-condensing
IP RatingIP54
IncludesLogger only (no accessories)
InterfaceIncludes SDI-12, Ethernet, MODBUS, RS232, RS422, RS485, USB
ModelDT80G-AL
Operating Temperature (Metric)-30°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Sensor TypesNumerous, including vibrating wire sensors, thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Weight (Metric)1.5 kg
Alarm TypesE-mail, SMS, Physical/Digital Output
Display TypeBacklit LCD, 2 line by 16 characters
TypeGeotechnical logger supporting vibrating wire strain gauges
Unit SizeEach