DataTaker DT82E Series Data Loggers
Thermo Fisher Scientific™
DataTaker DT82E Series Data Loggers
DataTaker is one of the world's leading brands of general purpose and specialised data loggers. DataTaker is supplier to broad range of customers across many industries including environmental, industrial, construction, manufacturing, process management, scientific, laboratory and education.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
DT82EMLogger with integrated modem and accessories
DT82ELogger and accessories
DT82E-ALLogger only (no accessories)
Catalog number DT82EM
Price (INR)
-
Includes:
Logger with integrated modem and accessories
The DataTaker DT82E series comprises a family of environmental data loggers, with two analog channels and many digital inputs. DataTaker measures and logs analog and digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, strain gauges, current loops, voltages, resistance and frequencies. DataTaker models may include an Ethernet port, Modem for remote access and SMS alerts, WiFi, MODBUS connection, and may include a built-in web server, dEX, for setup and for viewing/downloading of data.
The DataTaker DT82E series comprises a family of environmental data loggers, with two analog channels and many digital inputs.
The DataTaker DT82E series measures and logs analog & digital data from numerous electrical sensors or analog instruments, RTDs, thermocouples, temperature sensors, strain gauges, current loops, voltages, resistance and frequencies.
DataTaker DT82E models include an Ethernet port and a built-in web server, dEX for setup and for viewing/downloading of data. In addition:
  • DT82EM includes an integrated modem
Other Features:
  • Analog input channels set up as binary logic level inputs.
  • Digital inputs recording ON/OFF states of contacts, or counting pulses over a desired period.
  • Alarm channels and digital outputs to raise alarms (email / SMS / physical digital outputs) or control external processes.
  • Readings may be returned directly or via internal calculations based on a single channel or multiple channels.
  • Internal memory holds ∼10 million readings.
  • Connection to the host computer via Ethernet, USB, and serial Host Port.
  • Separate USB port for USB flash drives to be used for extra storage, or automatic unloading of data from the logger.
  • Built-in battery or auxiliary battery option.
  • Most data logging configurations are easily set up using the graphical web-based dEX interface and a standard web browser.
  • For more complex configurations, a script can be developed using the logger's own scripting language.
Specifications
Battery TypeNone, external power required
CalibrationNot Calibrated
Channels2 to 6 Analog, 8 Digital
Data Memory128 MB
Dimensions (L x W x H)180 x 137 x 65 mm
Humidity RangeMaximum: 85%, non-condensing
IP RatingIP54
IncludesLogger with integrated modem and accessories
InterfaceIncludes SDI-12, Ethernet, MODBUS (Slave)
ModelDT82EM
Operating Temperature (Metric)-45°C to +70°C (reduced battery life and LCD operation outside range -15°C to +50°C)
Sensor TypesNumerous, including thermocouples, thermistors, temperature sensors, strain gauges and bridge sensors
Weight (Metric)900 g
Alarm TypesE-mail, SMS, Physical/Digital Output
Display TypeBacklit LCD, 2 line by 16 characters
TypeLow Cost Environmental Datalogger
Unit SizeEach