DVG50 Digital Vacuum Gauge
Actual product may vary
Thermo Scientific™
DVG50 Digital Vacuum Gauge
Monitor performance and efficiency of vacuum systems with the Thermo Scientific™ DVG50 Digital Vacuum Gauge.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
DVG50-230230 V
DVG50-115115 V
Catalog number DVG50-230
Price (INR)
-
Voltage:
230 V
Monitor performance and efficiency of vacuum systems with the Thermo Scientific™ DVG50 Digital Vacuum Gauge. Vacuum levels can be used as a diagnostic tool to determine system integrity, to verify that sufficient vacuum is present (e.g., as required for lyophilization) or to monitor the progression of the concentration process.
  • Easy to read LED display
  • Chemical resistance for use with both high vacuum and oil-free pumps
  • Direct attachment between vacuum lines
Specifications
Description230V 50Hz; AC adapter required
Electrical Requirements230 V 50 Hz
Frequency50 Hz
Voltage230 V
TypeDigital Vacuum Gauge
Unit SizeEach