EnzChek™ Lipase Substrate, green fluorescent, 505/515
Invitrogen™

EnzChek™ Lipase Substrate, green fluorescent, 505/515

Green-fluorescent product

Have Questions?
Catalog NumberQuantity
E33955100 μg
Catalog number E33955
Price (INR)
-
Quantity:
100 μg

The nonfluorescent EnzChek™ lipase substrate, in the presence of lipases, produces a green-fluorescent product (excitation/emission maxima ∼507/515) for accurate lipase detection in solution.

Dissolve in high-quality anhydrous DMSO or chloroform to prepare a stock concentrations of up to 1 mM.

The nonfluorescent EnzChek™ lipase substrate, in the presence of lipases, produces a green-fluorescent product (excitation/emission maxima ∼507/515) for accurate lipase detection in solution.

Dissolve in high-quality anhydrous DMSO or chloroform to prepare a stock concentrations of up to 1 mM.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Excitation/Emission507/515 nm
For Use With (Application)Enzyme and Metabolic Activity
Molecular Weight (g/mol)1011.15 g/mol
Product LineEnzChek
Quantity100 μg
Shipping ConditionRoom Temperature
SubstrateLipase substrate
Detection MethodFluorescence
Emission Wavelength Range515 nm
Excitation Wavelength Range507 nm
FormSolid
Substrate PropertiesChemical Substrate
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C) and protect from light.

Citations & References (2)

Citations & References
Abstract
Stabilization of enzymes in silk films.
Authors:Lu S, Wang X, Lu Q, Hu X, Uppal N, Omenetto FG, Kaplan DL,
Journal:Biomacromolecules
PubMed ID:19323497
'Material systems are needed that promote stabilization of entrained molecules, such as enzymes or therapeutic proteins, without destroying their activity. We demonstrate that the unique structure of silk fibroin protein, when assembled into the solid state, establishes an environment that is conducive to the stabilization of entrained proteins. Enzymes (glucose ... More
Determination of lipoprotein lipase activity using a novel fluorescent lipase assay.
Authors:Basu D, Manjur J, Jin W,
Journal:J Lipid Res
PubMed ID:21270098
A novel, real-time, homogeneous fluorogenic lipoprotein lipase (LPL) assay was developed using a commercially available substrate, the EnzChek lipase substrate, which is solubilized in Zwittergent. The triglyceride analog substrate does not fluoresce, owing to apposition of fluorescent and fluorescent quenching groups at the sn-1 and sn-2 positions, respectively, fluorescence becoming ... More