Fastidious Anaerobe Broth HPA
Fastidious Anaerobe Broth HPA
Thermo Scientific™
Fastidious Anaerobe Broth HPA
Cultivate and enrich fastidious anaerobic bacteria from clinical, food, or environmental samples under anaerobic conditions.
Have Questions?
Catalog NumberFill Weight
EB0912H25.0 to 25.3 g
Catalog number EB0912H
Price (INR)
-
Request A Quote
Fill Weight:
25.0 to 25.3 g
Fastidious Anaerobe Broth HPA is used to cultivate and enrich fastidious anaerobic bacteria from clinical, food, or environmental samples under anaerobic conditions.
Specifications
ColorStraw/Green with or without thin resazurin layer
Fill Weight25.0 to 25.3 g
FormatUniversal bottle
Packaging Type100 Universal Bottles with screw cap closures in a box
Quantity100/Pk.
Shelf Life13 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C