Fastidious Anaerobe Broth with Glass Beads
Fastidious Anaerobe Broth with Glass Beads
Thermo Scientific™
Fastidious Anaerobe Broth with Glass Beads
Enrich and maintain anaerobic bacteria, where the glass beads assist in biofilm disruption or homogenizing viscous samples for improved microbial recovery and subculture.
Have Questions?
Catalog NumberFill Weight
EB1305C5.0 to 5.3 g
Catalog number EB1305C
Price (INR)
-
Request A Quote
Fill Weight:
5.0 to 5.3 g
Fastidious Anaerobe Broth with Glass Beads is used to enrich and maintain anaerobic bacteria, where the glass beads assist in biofilm disruption or homogenizing viscous samples for improved microbial recovery and subculture.
Specifications
ColorStraw/Green with or without a red resazurin layer and haemin particles
Fill Weight5.0 to 5.3 g
FormatUniversal bottle
Packaging Type100 Universal Bottles with screw cap closures in a box
Quantity100/Pk.
Shelf Life13 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C