Violet Red Bile Glucose Agar (ISO)
Thermo Scientific™

Violet Red Bile Glucose Agar (ISO)

Use this agar for the selective and differential isolation of Enterobacteriaceae
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill WeightFormat
EB1329F15.0 to 15.3 gUniversal Bottle
BO1329R198 to 202 g250 mL Sirop Bottle
BO1329M99.0-101.0g125 mL sirop bottle
Catalog number EB1329F
Price (INR)
-
Fill Weight:
15.0 to 15.3 g
Format:
Universal Bottle
Violet Red Bile Glucose Agar (ISO) is a selective differential medium for the isolation of Enterobacteriaceae
Specifications
ColorDark Purple
Fill Weight15.0 to 15.3 g
FormatUniversal Bottle
Packaging Type100 universal bottles with screw cap closure in a box
Quantity100/Pk.
Shelf Life52 weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2° to 25°C