InhibiSURE™ Viral Inactivation Medium
Thermo Scientific™

InhibiSURE™ Viral Inactivation Medium

Non-hazardous, inactivation media for viral collection and transport of SARS-CoV2.
Have Questions?
Catalog NumberQuantity
EB1359A100 Pk.
Catalog number EB1359A
Price (INR)
-
Quantity:
100 Pk.

Rapidly inactivate and stabilize viral RNA at ambient temperature with a non-hazardous formulation using Thermo Scientific™ InhibiSURE™ Viral Inactivation Medium. This liquid medium is intended for the collection, transportation, and inactivation of SARS-CoV-2.

Safety:

Reduce the risk of exposure during specimen collection, transport, and processing with InhibiSURE™ Viral Inactivation Medium. The medium rapidly inactivates viruses and stabilizes viral RNA at ambient temperature using a non-hazardous formula.

The device is intended to be used with unprocessed nasal, nasopharyngeal, and throat swab samples.

Easy to Use:

Maintain the integrity of nucleic acids with InhibiSURE™ Viral Inactivation Medium - a safe, novel, proprietary formula that destroys the viral envelope and disrupts capsid proteins, removing virulence.

Inactivation occurs within 30 minutes at room temperature (10-30°C). 

No further processing of the sample is required ahead of RNA extraction and PCR.

Product is CE marked but not 510(k)-cleared and not available for sale in the U.S. Availability of product in each country depends on local regulatory marketing authorization status.
For In Vitro Diagnostic Use.  
Specifications
Closure ColorPurple
CompatibilityInhibiSURE™ Viral Inactivation Medium has been designed to be compatible with nucleic acid purification and amplification workflows. It has been validated with the Thermo Scientific™ MagMax™ MVP II kit (product code A48383) and TaqPath™ COVID‑19 CE‑IVD RT‑PCR Kit (A48067)
DescriptionInhibiSURE™ Viral Inactivation Medium
For Use With (Application)Liquid medium intended to be used for the collection and inactivation of SARS-CoV-2.
Quantity100 Pk.
Storage Requirements2°C to 30°C
Volume (Metric)1.5 mL
Unit SizeEach