Eutech™ PC 700 Multi-parameter Meter
Eutech™ PC 700 Multi-parameter Meter
Eutech™ PC 700 Multi-parameter Meter
Thermo Scientific™

Eutech™ PC 700 Multi-parameter Meter

Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Have Questions?
Catalog NumberMeasurement Modes
ECPC70043SpH, Conductivity
Catalog number ECPC70043S
Price (INR)
104,896.00
Each
Measurement Modes:
pH, Conductivity
Price (INR)
104,896.00
Each
Your all-in-one solution for multi-parameter research work in the lab. The multi-parameter Eutech PC 700 measures pH, ORP, conductivity and temperature with a single meter. User-friendly with advanced features and a large custom-size display – perfect for research work in the lab.

Features

  • Large screen in a compact footprint
  • meter accepts pH and Redox electrodes, in addition to Conductivity/TDS – it's like having fourmeters in one!
  • Up to 5-point push button calibration
  • Selectable cell constant
  • Auto-ranging across 5 Conductivity ranges
  • Easy interchangeability of probes
  • Non-volatilememory holds up to 100 data points
  • Integral electrode holder

Applications

  • Water quality testing
  • Checkingmetal finishes
  • Solutions
  • Cooling tower water
  • Printing fountain solutions
  • Boiler water
  • Brines
  • Drillingmud
  • Rinse tanks
  • Ponds
  • Pollution control
  • Recirculating systems
  • Waste water
  • Industrial process systems
  • General use in laboratories
  • Titration
  • Quality assurance
  • Ecological studies
  • Food-processing

Specifications

Product Specification: Description

pH

  • Range: -2.00 to 16.00 pH
  • Resolution: 0.01 pH
  • Accuracy: ±0.01 pH
  • Cal. Points: Up to 5
  • Buffer Sets: USA (1.68, 4.01, 7.00, 10.01, 12.45), NIST (1.68, 4.01, 6.86, 9.18, 12.45)

ORP

  • Range: ±2000mV
  • Rel.mV Range: ±2000mV
  • Resolution: 0.1mV (±199.9mV) / 1mV (beyond)
  • Accuracy: ±0.2mV (±199.9mV) / ±2mV (beyond)

Conductivity

  • Range: 0.0 μS to 200.0mS
  • Resolution: 0.01 / 0.1 / 1 μS ; 0.01 / 0.1mS
  • Accuracy: ±1 % full scale
  • Cal. Points: Automatic (84 μS, 1413 μS, 12.88mS, 111.8mS);maximum 1 per range;manual (5 points);maximum 1 per range
  • Cell Constant: 0.1, 1.0, 10.0 (selectable)

TDS

  • Range: .. to 100.0 ppt @ 0.5 fact (200.0 ppt @ 1 factor)
  • Resolution: 0.01 / 0.1 / 1 ppm ; 0.01 / 0.1 ppt
  • Accuracy: ±1 % full scale
  • Cal. Points:manual (5 points);maximum 1 per range
  • TDS Factor: 0.40 to 1.00 (adjustable)

Temperature

  • Range (meter): 0.0 to 100.0 °C / 32.0 to 212.0 °F
  • Resolution: 0.1 °C / 0.1 °F
  • Accuracy: ±0.5 °C / ±0.9 °F
  • Coefficient (Per °C): 0.00 to 10.00 %
  • Normalization: 15.0 to 30.0 °C (adjustable)
  • Compensation Type & Range: Automatic with supplied cell ormanual, 0.0 to 100 °C / 32.0 to 212.0 °F, 0 to 80 °C /, 32.0 to 176.0 °F with supplied cell
  • Power Requirements: 9 V DC adapter, 1.3 A (100/240 VAC, SmPS)
Specifications
DescriptionEutech PC 700 Meter With pH Electrode (ECFC725211B), Conductivity/ATC Probe (CONSEN951D), Integral Electrode Holder & 1/24 VAC Adapter
IncludespH Electrode, Conductivity/ATC Probe
Resolution (pH)0.01
Display TypeLCD
Measurement ModespH, Conductivity
Range (Conductivity)0 μS/cm to 200 mS/cm
Range (TDS)100 ppt at 0.5 Factor, 200 ppt (at 1 Factor)
Temperature Range (Metric)0°C to 100°C
Unit SizeEach