Product Image
Thermo Scientific™
National™ Replacement Battery for Gen3 Electronic Crimpers and Decrimpers
For use with Thermo Scientific™ National Electronic Crimpers and Decrimpers.
Have Questions?
Catalog NumberFor Use With (Equipment)
ECR-CBATTGen3 Electronic Crimpers And Decrimper
Catalog number ECR-CBATT
Price (INR)
-
For Use With (Equipment):
Gen3 Electronic Crimpers And Decrimper
For use with Thermo Scientific™ National Electronic Crimpers and Decrimpers.
  • 6.4V Lithium Ion Battery
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionReplacement Battery
For Use With (Application)Industrial Chromatography
No. per Pack1 Ea.
Product LineNational
For Use With (Equipment)Gen3 Electronic Crimpers And Decrimper
Unit SizeEach