This power cord is appropriate for use with the GeneAmp™ PCR System 9700, Veriti™ thermal cyclers, and 2720 Thermal Cycler.Read more
Have Questions?
Catalog Number
Quantity
ED000651
1 each
Catalog number ED000651
Price (INR)
-
Quantity:
1 each
This power cord is appropriate for use with the GeneAmp™ PCR System 9700, Veriti™ thermal cyclers, and 2720 Thermal Cycler. This version is for use in European countries, excluding Switzerland, the United Kingdom, and Denmark.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)PCR Amplification
For Use With (Equipment)GeneAmp 9700, Veriti Thermal Cycler, 2720 Thermal Cycler