Cell Copper (Cu) Colorimetric Assay Kit
Invitrogen™

Cell Copper (Cu) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA00796 tests
Catalog number EEA007
Price (INR)
42,392.00
線上優惠
Ends: 31-Dec-2026
70,653.00
Save 28,261.00 (40%)
Each
Quantity:
96 tests
Price (INR)
42,392.00
線上優惠
Ends: 31-Dec-2026
70,653.00
Save 28,261.00 (40%)
Each

The Cell Copper Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of copper ion content in cells.

In this assay, under acidic conditions, copper ions in the sample react with a complexing agent to form a purple complex that has a maximum absorption peak at 580 nm. The copper ion content can be calculated indirectly by measuring the OD value at 580 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.18–5 μmol/L
Assay Sensitivity0.18 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time5 min.
Interassay CV3.1
Intraassay CV3
Quantity96 tests
Sample Volume100 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedCell Copper (Cu)
Total Assay Time10 min.
Detection MethodColorimetric
Sample TypeWhole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.