Cell Ferrous Iron Colorimetric Assay Kit
Invitrogen™

Cell Ferrous Iron Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA00896 tests
Catalog number EEA008
Price (INR)
47,691.00
Online offer
Ends: 31-Dec-2026
70,653.00
Save 22,962.00 (32%)
Each
Quantity:
96 tests
Price (INR)
47,691.00
Online offer
Ends: 31-Dec-2026
70,653.00
Save 22,962.00 (32%)
Each

The Cell Ferrous Iron Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ferrous ions in cells. Iron plays an important role in various physiological functions. The ferrous ion is a key element in heme and hemoglobin and is present in many biochemical reactions.

In this assay ferrous ions (Fe2+) in samples bind with probes in solution to form a complex that has a maximum absorption peak at 593 nm. The concentration of ferrous ions can be calculated indirectly by measuring the OD value.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.4–50 μmol/L
Assay Sensitivity0.4 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time10 min.
Interassay CV1.5
Intraassay CV1.3
Quantity96 tests
Sample Volume160 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedCell Ferrous Iron
Total Assay Time40 min.
Detection MethodColorimetric
Sample TypeWhole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.