H2S Colorimetric Assay Kit
H<sub>2</sub>S Colorimetric Assay Kit
Invitrogen™

H2S Colorimetric Assay Kit

The Hydrogen Sulfide (H2S) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement ofRead more
Have Questions?
Catalog NumberQuantity
EEA01196 tests
Catalog number EEA011
Price (INR)
47,691.00
Online offer
Ends: 31-Mar-2026
70,653.00
Save 22,962.00 (32%)
Each
Add to cart
Quantity:
96 tests
Price (INR)
47,691.00
Online offer
Ends: 31-Mar-2026
70,653.00
Save 22,962.00 (32%)
Each
Add to cart

The Hydrogen Sulfide (H2S) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of H2S in serum, plasma, and tissue samples. H2S is a crucial signal molecule involved in various biological processes, from cellular signal transduction to nerve conduction and protein synthesis, where it plays a role in regulating development, angiogenesis, and protecting organs from ischemia or reperfusion injury.

In this assay H2S reacts with an acetate solution to create soluble ZnS, where, in the presence of Fe3+, methylene blue is formed (which has a maximum absorption peak at 665 nm). The concentration of iron can be indirectly calculated by measuring the OD value.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

Specifications
Assay Range4.86–100 μmol/L
Assay Sensitivity2.75 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time20 min.
Interassay CV9.9
Intraassay CV3.1
Quantity96 tests
Sample Volume100 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedHydrogen Sulfide (H2S)
Total Assay Time80 min.
Detection MethodColorimetric
Sample TypePlasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.