Potassium (K) Turbidimetric Assay Kit
Invitrogen™

Potassium (K) Turbidimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA01896 tests
Catalog number EEA018
Price (INR)
42,392.00
Online offer
Ends: 31-Dec-2026
70,653.00
Save 28,261.00 (40%)
Each
Quantity:
96 tests
Price (INR)
42,392.00
Online offer
Ends: 31-Dec-2026
70,653.00
Save 28,261.00 (40%)
Each

The Potassium (K) Turbidimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of potassium content in serum, plasma, milk, tissue, cells, and other sample types. Potassium is an essential ion that plays a crucial role in various physiological processes, including muscle and nerve function, fluid balance, and cardiovascular health. Potassium deficiency and excess can each result in different signs and symptoms, including abnormal heart rhythm and various electrocardiographic abnormalities

In this assay, under alkaline conditions, potassium ions in the sample react with sodium tetraphenylborate to form potassium tetraphenylborate, a turbid white solution with little solubility. The turbidity is proportional to the potassium ion concentration, which can be calculated indirectly by measuring the OD value at 450 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.01–0.80 mmol/L
Assay Sensitivity0.002 mmol/L
For Use With (Equipment)Microplate Reader
Incubation Time5 min.
Interassay CV6.1
Intraassay CV1.1
Quantity96 tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedPotassium (K)
Total Assay Time20 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Milk, Plasma, Serum, Tissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.