Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (ABTS, enzyme method)
Invitrogen™

Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (ABTS, enzyme method)

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02396 tests
Catalog number EEA023
Price (INR)
-
Quantity:
96 tests

The Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (ABTS, enzyme method) is a highly reliable and sensitive assay designed for the quantitative measurement of T-AOC in serum, plasma, cells, and other biological sample types. T-AOC refers to the overall ability of a biological sample to counteract oxidative stress by neutralizing reactive oxygen species (ROS) and other harmful free radicals. It represents the cumulative effect of both enzymatic and non-enzymatic antioxidants present in the sample.

The assay uses ABTS, which is oxidized to ABTS+ in solution. ABTS+ is measured at 414 or 734 nm and is proportional to the T-AOC in the sample.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

Specifications
Assay Range0.047–1.50 mmol/L
Assay Sensitivity0.047 mmol/L
For Use With (Equipment)Microplate Reader
Incubation Time6 min.
Interassay CV4.1
Intraassay CV2.2
Quantity96 tests
Sample Volume10 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedTotal Antioxidant Capacity (T-AOC)
Total Assay Time20 min.
Detection MethodColorimetric
Sample TypePlasma, Saliva, Serum, Tissue Homogenates, Urine, Whole Cells
Unit SizeEach
Contents & Storage
Reagent 5: –20°C
Other reagents: 2–8°C