Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (FRAP method)
Invitrogen™

Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (FRAP method)

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02496 tests
Catalog number EEA024
Price (INR)
47,691.00
Online offer
Ends: 31-Dec-2026
70,653.00
Save 22,962.00 (32%)
Each
Quantity:
96 tests
Price (INR)
47,691.00
Online offer
Ends: 31-Dec-2026
70,653.00
Save 22,962.00 (32%)
Each

The Total Antioxidant Capacity (T-AOC) Colorimetric Assay Kit (FRAP method) is a highly reliable and sensitive assay designed for the quantitative measurement of T-AOC in serum, plasma, tissues, cells, cell culture supernatant, saliva, and urine sample types. T-AOC refers to the overall ability of a biological sample to counteract oxidative stress by neutralizing reactive oxygen species (ROS) and other harmful free radicals. It represents the cumulative effect of both enzymatic and non-enzymatic antioxidants present in the sample.

The assay quantifies the ability of the sample to reduce ferric ions (Fe3+) to ferrous ions (Fe2+) using the ferric reducing antioxidant power (FRAP) method under acidic conditions. T-AOC can be measured using the absorbance value of the resulting blue solution at 593 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.23–2 mmol Trolox Equiv./L
Assay Sensitivity0.049 mmol/L
For Use With (Equipment)Microplate Reader
Incubation Time3-5 min.
Interassay CV8.1
Intraassay CV3.9
Quantity96 tests
Sample Volume5 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedTotal Antioxidant Capacity (T-AOC)
Total Assay Time20 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Saliva, Serum, Tissue Homogenates, Urine, Whole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.